This product is now discontinued, View Related Products

CoorsTek Spatula/Spoons, Porcelain

Spatula/spoons are glazed all over. Autoclavable.
17,300.00INR / Pkg of 1

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Overall Length (in)
Availability Pricing
CoorsTek 60477 Glazed Porcelain Spoon/Spatula, 0.1 mL, 3-7/8" L; 1/Pk WW-17828-00 Mfr # 60477
COMPARE
0.1 3 7/8
Call For Price
CoorsTek 60478 Glazed Porcelain Spoon/Spatula, 0.3 mL, 4-7/8" L; 1/Pk WW-17828-02 Mfr # 60478
COMPARE
0.3 4 7/8
Call For Price
CoorsTek 60480 Glazed Porcelain Spoon/Spatula, 0.5 mL, 6-1/2" L; 1/Pk WW-17828-04 Mfr # 60480
COMPARE
0.5 6 1/2
Call For Price
CoorsTek 60481 Glazed Porcelain Spoon/Spatula, 1.0 mL, 7-3/4" L; 1/Pk WW-17828-06 Mfr # 60481
COMPARE
1 7 3/4
Call For Price
CoorsTek 60483 Spatula/spoon, glazed porcelain, autoclavable, , 6.0 mL, 10" (254 mm) L WW-17828-08 Mfr # 60483
COMPARE
6 10
₹17,300.00INR / Pkg of 1
CoorsTek 60477 Spatula/spoon, glazed porcelain, autoclavable, , 0.1 mL, 3-7/8" (99 mm) L, cs of 24 WW-17828-20 Mfr # 60477
COMPARE
0.1 3 7/8
Call For Price
CoorsTek 60478 Spatula/spoon, glazed porcelain, autoclavable, , 0.3 mL, 4-7/8" (123 mm) L, cs of 24 WW-17828-22 Mfr # 60478
COMPARE
0.3 4 7/8
Call For Price
CoorsTek 60480 Spatula/spoon, glazed porcelain, autoclavable, , 0.5 mL, 6-1/2" (165 mm) L, cs of 12 WW-17828-24 Mfr # 60480
COMPARE
0.5 6 1/2
Call For Price
CoorsTek 60481 Spatula/spoon, glazed porcelain, autoclavable, , 1.0 mL, 7-3/4" (195 mm) L, cs of 12 WW-17828-26 Mfr # 60481
COMPARE
1 7 3/4
Call For Price
CoorsTek 60483 Spatula/spoon, glazed porcelain, autoclavable, , 6.0 mL, 10" (254 mm) L, cs of 8 WW-17828-28 Mfr # 60483
COMPARE
6 10
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat