This product is now discontinued, View Related Products
CoorsTek Mortars, Porcelain
Mortars are glazed all over except for grinding surfaces and the outside bottom. Features include lip and pour spout. Mortar and pestle are sold as separate pieces. Autoclavable.
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Mortars Capacity (mL) Mortars Height (mm) Mortars Top OD (mm)
Availability Pricing
CoorsTek 66310 Porcelain Mortar, 50 mL, pack of 1 WW-17820-00 Mfr # 66310
COMPARE
50 47 70
Call For Price
CoorsTek 66313 Porcelain Mortar, 65 mL, pack of 1 WW-17820-02 Mfr # 66313
COMPARE
65 53 80
Call For Price
CoorsTek 66316 Porcelain Mortar, 145 mL, pack of 1 WW-17820-04 Mfr # 66316
COMPARE
145 70 90
Call For Price
CoorsTek 66319 Porcelain Mortar, 275 mL, pack of 1 WW-17820-06 Mfr # 66319
COMPARE
275 70 115
Call For Price
CoorsTek 66322 Porcelain Mortar, 400 mL, pack of 1 WW-17820-08 Mfr # 66322
COMPARE
400 80 130
Call For Price
CoorsTek 66325 Porcelain Mortar, 750 mL, pack of 1 WW-17820-10 Mfr # 66325
COMPARE
750 110 163
Call For Price
CoorsTek 66328 Porcelain Mortar, 1900 mL, pack of 1 WW-17820-12 Mfr # 66328
COMPARE
1900 130 210
Call For Price
CoorsTek 60313 Porcelain Mortar, 65 mL, case of 18 WW-17820-22 Mfr # 66313
COMPARE
65 53 80
Call For Price
CoorsTek 60316 Porcelain Mortar, 145 mL, case of 12 WW-17820-24 Mfr # 66316
COMPARE
150 65 85
Call For Price
CoorsTek 60319 Porcelain Mortar, 275 mL, case of 8 WW-17820-26 Mfr # 66319
COMPARE
300 75 105
Call For Price
CoorsTek 60322 Porcelain Mortar, 400 mL, case of 8 WW-17820-28 Mfr # 66322
COMPARE
420 88 120
Call For Price
CoorsTek 66325 Porcelain Mortar, 750 mL; 4/CS WW-17820-30 Mfr # 66325
COMPARE
750 110 163
Call For Price
CoorsTek 66328 Porcelain Mortar, 1900 mL, case of 2 WW-17820-32 Mfr # 66328
COMPARE
1900 130 210
Call For Price
CoorsTek 66331 Porcelain Mortar, 4000 mL, case of 1 WW-17820-34 Mfr # 66331
COMPARE
4000 150 283
Call For Price
Showing 1- 14 of 14