CoorsTek High-Form Crucibles, High Alumina
Manufactured from 99.8% pure aluminum oxide, these high-form crucibles are highly resistant to chemicals, alkalines, and other fluxes. Crucible is inactive in hydrogen and carbonaceous atmospheres. Withstands temperatures to 3182°F (1750°C). Crucible covers sold separately. Autoclavable.
9,600.00 - ₹32,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
CoorsTek 65501 High-Form Crucible, High Alumina; 10 mL WW-17804-01 Mfr # 65501
COMPARE
31 10 29
₹9,600.00INR / Each
CoorsTek 65503 High-Form Crucible, High Alumina; 20 mL WW-17804-03 Mfr # 65503
COMPARE
35 20 38
₹11,400.00INR / Each
CoorsTek 65504 High-Form Crucible, High Alumina; 50 mL WW-17804-05 Mfr # 65504
COMPARE
48 50 48
₹14,700.00INR / Each
CoorsTek 65505 High-Form Crucible, High Alumina; 100 mL WW-17804-07 Mfr # 65505
COMPARE
58 100 58
₹20,200.00INR / Each
CoorsTek 65506 High-Form Crucible, High Alumina; 250 mL WW-17804-09 Mfr # 65506
COMPARE
93 250 76
₹32,800.00INR / Each