This product is now discontinued, View Related Products
Convertible Utility Carts
Convert from a box-top to a flat-top quickly and easily
Hinged side gates—customize cart configurations in a snap. HDPE construction with I-beam design—each shelf supports up to 200 pounds (90.0 kg). Easily load and unload oversized boxes. includes 5" ball bearing wheels—rear wheels lock for stabilization. Molded-in compartments keep supplies accessible. Optimize organization and storage space with Akro-Bins® used in rail hanging system (sold separately) Rail Hanging System includes a durable panel with nine hanging rails. Hang bins and modular cabinets on the rails to organize and store small parts. Akro-Bins® sold on pages AkroBin—Reg TM Myers Industries, Inc.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Number of Shelves Shelf Height (in)
Availability Pricing
Akro-Mils 30930,BEIGE Convertible Utility Cart, 2-Shelf, Small, 19.5" x 40", 200 lbs Cap WW-47153-10 Mfr # 30930,GRAY
COMPARE
200 2
Call For Price
Akro-Mils 30936,BEIGE Convertible Utility Cart, 2-Shelf, Large, 24" x 45", 200 lbs Cap WW-47153-12 Mfr # 30936 GRAY
COMPARE
200 2