Cole-Parmer® Continuous Flow Cell
Recommended for the treatment of low-viscosity samples where the required insonation time is short
  • Made of autoclavable stainless steel
This low-volume cell screws onto the standard 2" dia probe and forces all of the sample solution to pass across the probe face. Use the cell to emulsify, disperse, and homogenize samples at flow rates up to 5 GPH (19 liters/hour). Fittings accept 16" ID soft tubing.
129,000.00 - ₹232,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Ultrasonic Processor Continuous Medium Flow Cell
₹232,000.00INR / Each
COMPARE
Replacement probe for Ultrasonic Processor Continuous Medium Flow Cell
₹129,000.00INR / Each