PYREX® Kuderna-Danish Concentrator Apparatus with Hooks
Efficiently boil away pestistides for accurate testing
  • Held together using glass hooks and two 1 1/4 inch stainless steel springs
Kuderna-Danish Concentrators boil away samples to allow testing for pesticides or other pollutants prior to instrumental analysis. Include three-ball Snyder column with venting dimples and graduated collection tube. Available with springs mounted on collection tube.
102,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Range
Availability Pricing
Corning 2158-500 Concentrator with Springs, 500 Ml WW-34530-00 Mfr # 2158-500
COMPARE
500 mL
₹102,000.00INR / Each