This product is now discontinued, View Related Products
Combs for Cole-Parmer High-Throughput Horizontal Gel System
Optimize performance of your High-Throughput Horizontal Electrophoresis Gel System
These combs are compatible with your Cole-Parmer 28562-00 system. Combs are single-sided and height-adjustable; available in three thicknesses to accommodate different sample volumes. The 27- and 28-well combs are spaced to accept samples loaded using multi-channel pipettors. The 27-well combs include three molecular weight marker lanes with two spacings of 12 sample lanes; 28-well combs include four molecular weight marker lanes with three spacings of eight sample lanes.
Call For Price
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Comb for High-Throughput Horizontal Gel System; 26 Wells, 1.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 27 Wells, 1.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 28 Wells, 1.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 51 Wells, 1.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 102 Wells, 1.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 26 Wells, 1.5 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 27 Wells, 1.5 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 28 Wells, 1.5 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 51 Wells, 1.5 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 102 Wells, 1.5 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 26 Wells, 2.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 27 Wells, 2.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 28 Wells, 2.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 51 Wells, 2.0 mm Thick
Call For Price
COMPARE
Comb for High-Throughput Horizontal Gel System; 102 Wells, 2.0 mm Thick
Call For Price
Showing 1- 15 of 15