Cole-Parmer® Essentials Wide-Mouth Plastic Bottles, PP
Wide-mouth opening provides easier filling and sampling
  • Leakproof and chemical resistance
  • FDA 21 CFR compliant for food contact
  • Free of heavy metals and RoHS compliant
  • BPA and Phthalate free
  • Approved for use from -40 to 250°F (-40 to 121°C)
  • Autoclavable
Manufactured from USP Class VI virgin polypropylene (PP) resin, these wide mouth bottles offer excellent chemical resistance and clarity. Precision molded for consistent wall thickness and smooth interior and exterior sufaces. The wide mouth opening is perfect for sample collection and allows easy filling and emptying of solids, powders, and viscous liquids.
3,400.00 - ₹22,900.00INR / Case of 24
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28 mm Polypropylene(PP)
₹17,100.00INR / Case of 72
COMPARE
30 28 mm Polypropylene(PP)
₹4,600.00INR / Pkg of 12
COMPARE
1000 63 mm Polypropylene(PP)
₹22,900.00INR / Case of 24
COMPARE
60 28 mm Polypropylene(PP)
₹3,400.00INR / Pkg of 12
COMPARE
125 38 mm Polypropylene(PP)
₹4,600.00INR / Pkg of 12
COMPARE
250 43 mm Polypropylene(PP)
₹9,000.00INR / Pkg of 12
COMPARE
500 53 mm Polypropylene(PP)
₹12,700.00INR / Pkg of 12
COMPARE
1000 63 mm Polypropylene(PP)
₹9,600.00INR / Pkg of 6
COMPARE
2000 100 mm Polypropylene(PP)
₹3,600.00INR / Each
COMPARE
4000 100 mm Polypropylene(PP)
₹4,800.00INR / Each
Showing 1- 10 of 10