This product is now discontinued, View Related Products
Spex® VapLock™ Silicone Tubing for Solenoid Pinch Valves
Durable tubing withstands repeated pinch valve usage
  • Complies with USP XXII, Class VI, FDA, and USDA
  • FDA master file listed (MAF 819)
  • Sterilized through steam autoclaving, boiling, dry heat, and gamma irradiation
Designed to withstand rigorous usage with most pinch valve applications, the platinum-cured silicone tubing is ideal for high-purity applications. Rated for 500,000 pinch valve cycles.
The tubing features a low compression set and resists sticking shut even after prolonged
periods in the closed position. Excellent memory characteristics ensure full opening and unencumbered flow.
36,500.00 - ₹59,600.00INR / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
25 1/32 3/32
Call For Price
COMPARE
25 1/16 1/8
₹36,500.00INR / Pkg of 1
COMPARE
25 1/16 3/16
Call For Price
COMPARE
20 1/8 1/4
₹59,600.00INR / Pkg of 1