This product is now discontinued, View Related Products
Cole-Parmer® VapLock™ NPT(M) Threaded Adapters
Designed for permanent connection
  • Perfect for general-purpose and single-use applications, to connect tubing with rest of system
  • Natural polypropylene material offers very good chemical resistance
  • Hex nut construction – use in high-pressure applications
  • Sterilize via autoclave or ethylene oxide (EtO)
The VapLock barbed to threaded fittings are made of durable polypropylene for harsh applications. Polypropylene is compatible with most chemicals and is easy to sterilize via autoclave or ethylene oxide. The angled barb design ensures compatibility with different types of tubing. The hex nut construction provides a tighter, leakproof seal while expanding the inner tubing by 50%, allowing for use in high-pressure applications. The design ensures that the tubing is easy to attach to the fitting, but difficult to remove.
170.00 - ₹180.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem Availability Pricing
Spex VapLock™ Polypropylene, Straight, Hose Barb to Threaded Adapter, 3/32" ID x 1/16-27 NPT WW-12024-04 Mfr # 16220-6005
COMPARE
₹170.00INR / Each
Spex VapLock™ Polypropylene, Straight, Hose Barb to Threaded Adapter, 1/16" ID x 1/8-27 NPT WW-12024-08 Mfr # 18210-6
COMPARE
₹180.00INR / Each
Spex VapLock™ Polypropylene, Straight, Hose Barb to Threaded Adapter, 3/32" ID x 1/8-27 NPT WW-12024-10 Mfr # 18220-6