This product is now discontinued, View Related Products
Cole-Parmer Vacuum Filtration System
Pre-sterilized filtration systems for every application
  • Gamma sterilized to reduce the risk of contamination
  • Made with a low protein binding and surfactant-free PES membrane
  • Available in two membrane pore size options for an assortment of applications
The vacuum filtration systems include the upper funnel and dust cap along with a media bottle. The design features an ergonomic bottle shape with molded finger grips. The funnel and bottle are made of polystyrene material. These systems are available in three different capacities: 250 mL, 500 mL, and 1000 mL. These capacities are available in two pore size options. The 0.1 µm pore size is mycoplasma filter grade and the 0.45 µm filter is clarification grade.
These systems can be used in conjunction with the optional pedestal or cradle ring to create a hands-free filtration system.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Membrane Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
1000 90 0.1
Call For Price
COMPARE
1000 90 0.45
Call For Price
COMPARE
250 66 0.1
Call For Price
COMPARE
250 66 0.45
Call For Price
COMPARE
500 90 0.1
Call For Price
COMPARE
500 90 0.45