This product is now discontinued, View Related Products
Cole-Parmer USB Data Acquisition Modules
No external power connection required—powered from the USB port
  • Ideal for process, manufacturing, quality control, testing, or lab
  • Fully compatible with USB 1.1 and USB 2.0
  • Software suite included is supported by TracerDaq™ Pro, DASYLab®, LabVIEW™, and is compatible with most Microsoft Windows® operating systems
These economical plug and play data acquisition modules are ideal for monitoring process, manufacturing, quality control, testing, or lab environments. Units feature multifunction I/O, digital I/O, temperature measurement capabilities, and integrated screw terminals - eliminating the need for external cable connections. The external green LED confirms communication to your PC. The rugged compact design is perfect for harsh environments and for saving valuable benchspace. All modules are fully compatible with USB 1.1 as well as USB 2.0, and are powered by the 5 volt USB supply from your computer.
5 PRODUCT OPTIONS
Compareitem specifications
Data Sample Rate (MHz) Number Of Analog Differential Channels Number Of Analog Output Channels
Availability Pricing
COMPARE
1.2k Samples/second 4 2
Call For Price
COMPARE
50k Samples/second 4 2
Call For Price
COMPARE
50k Samples/second
Call For Price
COMPARE
2 Samples/second per channel 8
Call For Price
COMPARE
2 Samples/second per channel 8