Environmental Express® Uncertified VOA Vials
Borosilicate glass vials meet EPA guidelines for contamination-free containers
  • Choose between 40- and 60-mL capacities
  • Include standard screw-top caps
Level 3 VOA vials are not certified and ideal for general use containers that meet EPA recommended guidlines. Vials come with closed-top caps with PTFE liner. Choose from clear vials to easily view samples or amber vials to protect light-sensitive samples.
11,500.00 - ₹26,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Color Volume (mL)
Availability Pricing
COMPARE
Clear 10000
₹11,500.00INR / Each
COMPARE
Amber VOA Vials, 60 mL, Open-Top Cap; 72/Cs Amber 60
₹19,100.00INR / Case of 12
COMPARE
Clear VOA Vials, 40 mL, Closed-Top Cap, Economy Case; 144/Cs Clear 40
₹19,900.00INR / Case of 66
COMPARE
Vial, Amber Econo CT PTFE Liner, 60mL, 144/C Amber 60
₹26,100.00INR / Case of 144
COMPARE
Vial, Amber Econo OT, 60mL, Spt, 144/C Amber 60
₹24,100.00INR / Case of 144
COMPARE
Vial, 24mm Amber OT, 60mL, 72/C Amber 60
₹14,600.00INR / Case of 72
COMPARE
Vial, Clear OT Econo, 60mL, 144/C Clear 60
₹23,500.00INR / Case of 144
COMPARE
Vial, 24mm Amber CT, 60mL, 72/C White 60
₹18,500.00INR / Case of 72
Showing 1- 8 of 8