This product is now discontinued, View Related Products

Cole-Parmer UN/DOT Solvent Waste Systems with Rigid Tubing Ports

Provide safe HPLC waste disposal in your lab
  • Certified for UN/DOT Packaging Group II for hazardous chemicals
  • Carbon filter captures potentially harmful vapors
  • Multiple ports adapt cap to any HPLC system
  • Filter adapter elbow provides storage flexibility, especially for tight lab spaces
This solvent waste system allows easy and safe disposal of HPLC waste in the lab. The system includes a UN/DOT container with two caps; one with ports for waste collection and one to seal for transportation. Caps come with a variety of port sizes to be used with complex HPLC systems. When not in use, the ports can be plugged.
Call For Price

20 variations of this product are available.

See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Cap / Stopper Size Special Features
Availability Pricing
COMPARE
13.5 70 mm 6 x 1/16\" OD Tubing
Call For Price
COMPARE
13.5 70 mm 6 x 1/8\" OD Tubing
Call For Price
COMPARE
13.5 70 mm 4 x 1/8\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
13.5 70 mm 4 x 1/16\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
13.5 70 mm 4 x 1/8\" and 4 x 1/4\" OD Tubing
Call For Price
COMPARE
13.5 70 mm 4 x 1/16\" and 4 x 1/4\" OD Tubing
Call For Price
COMPARE
20 70 mm 6 x 1/16\" OD Tubing
Call For Price
COMPARE
20 70 mm 6 x 1/8\" OD Tubing
Call For Price
COMPARE
20 70 mm 4 x 1/8\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
20 70 mm 4 x 1/16\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
20 70 mm 4 x 1/8\" and 4 x 1/4\" OD Tubing
Call For Price
COMPARE
20 70 mm 4 x 1/16\" and 4 x 1/4\" OD Tubing
Call For Price
COMPARE
10 51 mm 6 x 1/16\" OD Tubing
Call For Price
COMPARE
10 51 mm 6 x 1/8\" OD Tubing
Call For Price
COMPARE
60 70 mm 6 x 1/16\" OD Tubing
Call For Price
COMPARE
60 70 mm 6 x 1/8\" OD Tubing
Call For Price
COMPARE
60 70 mm 4 x 1/8\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
60 70 mm 4 x 1/16\" and 3 x 1/4\" OD Tubing
Call For Price
COMPARE
60 70 mm 4 x 1/8\" and 4 x 1/4\" OD Tubing
Call For Price
COMPARE
60 70 mm 4 x 1/16\" and 4 x 1/4\" OD Tubing
Call For Price
Showing 1- 20 of 20

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat