This product is now discontinued, View Related Products

Cole-Parmer UN/DOT Solvent Waste System Carboy Caps

Collect hazardous waste safely
  • UN/DOT approved cap suitable for packing groups II and III
  • Activated carbon exhaust filter traps solvent vapors
  • Included elbow adapter saves vertical space
When paired with a UN/DOT-approved container with the same rating*, these carboy cap assemblies create a safe system for chemical waste collection and disposal. Fittings secure waste lines to cap, providing a closed loop to prevent harmful fumes from escaping. Activated carbon exhaust filter adsorbs vapors, allowing only clean air to return to the lab environment. Polypropylene cap with sulfur-cured EPDM gasket and fittings made of acetal and ETFE are chemically resistant to most HPLC-grade solvents.
Call For Price

30 variations of this product are available.

See All Product Options Below
30 PRODUCT OPTIONS
Compareitem specifications
Cap / Stopper Size Special Features
Availability Pricing
COMPARE
51 mm 6 x 1/8\" OD tubing
Call For Price
COMPARE
51 mm 6 x 1/16\" OD tubing
Call For Price
COMPARE
70 mm 6 x 1/16\" OD tubing
Call For Price
COMPARE
70 mm 6 x 1/8\" OD tubing
Call For Price
COMPARE
70 mm 6 x 1/16\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 6 x 1/8\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/8\" & 3 x 1/4\" OD tubing
Call For Price
COMPARE
70 mm 4 x 1/16\" & 3 x 1/4\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/8\" & 3 x 1/4\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/8\" & 4 x 1/4\" OD tubing
Call For Price
COMPARE
70 mm 4 x 1/16\" & 1 x 1/4\" OD tubing, 3 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/8\" & 1 x 1/4\" OD tubing, 3 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/8\" OD tubing, 4 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/16\" & 1 x 1/4\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 4 x 1/16\" and 4 x 1/4\" OD tubing
Call For Price
COMPARE
70 mm 4 x 1/16\" OD tubing, 4 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 6 x 1/16\" OD tubing
Call For Price
COMPARE
70 mm 6 x 1/8\" OD tubing
Call For Price
COMPARE
70 mm 6 x 1/16\" OD tbing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
COMPARE
70 mm 6 x 1/8\" OD tubing, 1 x 1/4\" or 3/8\" hose barb
Call For Price
Showing 1 - 20 of 30

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat