This product is now discontinued, View Related Products
Cole-Parmer Ultra-Durable Nitrile Gloves
Tough without compromising comfort and grip
  • Tear resistance
  • Exceptional durability
  • Superior chemical resistance
  • Ambidextrous with comfort fit for low fatigue
  • Low particulates
  • Free from allergenic protein
Manufactured using an innovative and exclusive nitrile formulation that provides outstanding chemical and mechanical properties. Offers comfort and dexterity combined with protection and strength.
Light weight, yet safe and durable. Fully textured surface finish offers confident grip and tactility for fine work. Rolled cuff provides added protection at wrist and ensures secure fit. Ideal choice for environments prone to static.
Applications include: pharmaceuticals, paint shops, laboratory analysis. general manufacturing, and e.lectronics
Complies with ASTM D6319, D6124, EN455.1, EN455.2, ISO 9001, and ISO 2859.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (in) Material Size
Availability Pricing
COMPARE
9 1/2 Nitrile Small
Call For Price
COMPARE
9 1/2 Nitrile Medium
Call For Price
COMPARE
9 1/2 Nitrile Large
Call For Price
COMPARE
9 1/2 Nitrile X-Large
Call For Price
COMPARE
9 1/2 Nitrile Small
Call For Price
COMPARE
9 1/2 Nitrile Medium
Call For Price
COMPARE
9 1/2 Nitrile Large
Call For Price
COMPARE
9 1/2 Nitrile X-Large
Call For Price
Showing 1- 8 of 8