Cole-Parmer Twist-Lock Conductivity Sensors
Ideal for by-pass streams, sample systems, and flow through applications
  • Ten-foot cables with 4-pin Military detachable connector
  • Built-in 100 Pt RTD
Rugged twist-lock conductivity cells feature a 12-mm sensor and automatic temperature compensation element permanently sealed in a Ryton®PPS housing. Two external Viton® O-rings protect against leakage, and locking pins ensure secure positioning. Ribbed-barrel design provides easy handling. The 1" NPT(M) top thread allows for attachment of conduit.
Use the Twist-Lock sensor with adapter (listed below) for conversion of a standard pipe tee to a flow through sensor aqssembly. The optional cable provides an additional 20 feet (6 meters) of coverage.
49,700.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Conductivity Cell, twist-lock, K=0.1
₹49,700.00INR / Each