This product is now discontinued, View Related Products
Cole-Parmer Turbine Mixing Head
Mix with low surface turbulence and high energy efficiency
  • No external blades, reduces risk of injury during cleaning
  • Made from high quality AISI 316 Stainless Steel
  • Unique design produces “full mix” with no dead spots
  • Anaerobic heads mix without air bubbles
  • Aerobic heads mix air into solutions
These mixing heads are part of the Teras Turbin mixer assembly for uniform and homogenous mixing results inside closed containers.
Match different mixing heads, shafts, and cap assemblies to create one of four mixing styles. Order a cap mixer assembly, shaft, mixing head, and connector to create a complete mixer system.
Call For Price
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Max Volume (Liters) Description
Availability Pricing
COMPARE
5 Turbine Mixing Head, open bottom, anaerobic, 15 mm
Call For Price
COMPARE
15 Turbine Mixing Head, open bottom, anaerobic, 25 mm
Call For Price
COMPARE
15 Turbine Mixing Head, open bottom, aerobic/full mix, 25 mm
Call For Price
COMPARE
60 Turbine Mixing Head, open bottom, anaerobic, 50 mm
Call For Price
COMPARE
60 Turbine Mixing Head, small inlet bottom, anaerobic, 50 mm
Call For Price
COMPARE
60 Turbine Mixing Head, open bottom, aerobic/full mix, 50 mm
Call For Price
COMPARE
60 Turbine Mixing Head, small inlet bottom, aerobic/full mix, 50 mm
Call For Price
COMPARE
100 Turbine Mixing Head, open bottom, anaerobic, 100 mm
Call For Price
COMPARE
100 Turbine Mixing Head, small inlet bottom, anaerobic, 100 mm
Call For Price
COMPARE
100 Turbine Mixing Head, open bottom, aerobic/full mix, 100 mm
Call For Price
COMPARE
100 Turbine Mixing Head, small inlet bottom, aerobic/full mix, 100 mm
Call For Price
Showing 1- 11 of 11