This product is now discontinued, View Related Products
Cole-Parmer Tube Rotator
Unique rotisserie system for mixing horizontally, vertically, or any position in between
  • Changing the mixing angle is easy — simply turn the rotisserie
  • Acheive 360° of rotation and a mixing angle range from 0 to 90°
Shaker includes a 36 x 1.5/2.0 ml rotisserie. Interchangeable rotisseries (sold separately below) accept tubes from 1.5 to 50 ml. Each rotisserie is split into two halves, so the halves can be exchanged to mix more than one tube type at a time. In addition, each rotisserie half can be rotated independently from a center point on the shaft. Ideal for a variety of different laboratory settings such as Molecular Biology, Biochemistry, research and Histochemistry. Small enough to fit on a crowded bench, cold room, incubator, or biological hood.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
Call For Price
COMPARE
230