Cole-Parmer® Essentials Trace Metal Analysis Pipette Tips
Tips are certified trace-metal free to prevent sample contamination
  • Constructed of virgin polypropylene
  • All materials used in the manufacture of these tips are free of trace metals
These universal pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit to variety of pipettors to ensure laboratory standardization. A reference mark at 10 µL lets you check pipettor calibration and visually estimate sample size. Tips are packaged in racks for easy access. Tips are tested by an independent laboratory and certified to be RNAse- and DNAse- free, a trace metal certificate is available upon request.
14,400.00 - ₹19,500.00INR / Pkg of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Package Of
Availability Pricing
COMPARE
1000 101 10 Racks of 100 Tips
₹19,500.00INR / Pkg of 1000
COMPARE
200 1 10 Racks of 96 Tips
₹14,400.00INR / Pkg of 960