This product is now discontinued, View Related Products
Cole-Parmer® Tall-Form Berzelius Beakers
Save space with tall-form beakers
  • Borosilicate glass construction with pour spout
These tall-form Berzelius beakers feature uniform wall thickness and a flat bottom making them ideal for heat applications Beakers have white, double-scale, metric graduations for easy measurment identification. The suggested maximum temperature is 460°F (240°C) for normal use, and 750°F (400°C) for short-term exposure.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material
Availability Pricing
COMPARE
150 Borosilicate Glass
Call For Price
COMPARE
250 Borosilicate Glass
Call For Price
COMPARE
400 Borosilicate Glass
Call For Price
COMPARE
600 Borosilicate Glass
Call For Price
COMPARE
800 Borosilicate Glass