Cole-Parmer® Submersion Housings
Protective housings for 12-mm electrodes
  • Protective housings provide an economical alternative
This 41''L x 1-1/16'' OD pipe is welded to a PVC connection box, and sealed with VITON O-rings for superior protection. Bottom tube detaches for electrode installation. CPVC model withstands temperatures up to 176°>F (80°>C); KYNAR model up to 230°>F (110°>C). Order extension cables for both models separately.
23,600.00 - ₹54,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrode Housing, submersible, CPVC w/ Viton o-rings
₹23,600.00INR / Each
COMPARE
pH Electrode Housing, submersible, PVDF w/ Viton o-rings
₹54,100.00INR / Each