Cole-Parmer® Sterile Serological Pipettes, PS
Easily recognize pipette capacity—color-coded stripes and matching packaging
  • Virgin polystyrene is gamma irradiated for sterility
  • Available bulk bagged or individually wrapped and bulk packed in bags
  • Premium material barrier plug
These sterile serological pipettes feature bold-colored positive and negative graduations making them easy to read while flling or dispensing. Color coded stripes and packaging enable quick volume identification.
All serological pipettes are manufactured to the ASTM Standard Specification for Serological Pipettes E-934.
12,900.00 - ₹65,600.00INR / Case of 200
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (mL) Package Of
Availability Pricing
COMPARE
Yellow 1 100 individually wrapped per bag
₹34,800.00INR / Case of 800
COMPARE
Green 2 100 individually wrapped per bag
₹34,300.00INR / Case of 600
COMPARE
Blue 5 50 individually wrapped per bag
₹12,900.00INR / Case of 200
COMPARE
Purple 50 30 individually wrapped per bag
₹34,800.00INR / Case of 90
COMPARE
Yellow 1 50 bulk wrapped per bag
₹40,100.00INR / Case of 1000
COMPARE
Green 2 50 bulk wrapped per bag
₹40,300.00INR / Case of 800
COMPARE
Blue 5 25 bulk wrapped per bag
₹29,100.00INR / Case of 500
COMPARE
Orange 10 25 bulk wrapped per bag
₹32,200.00INR / Case of 500
COMPARE
Orange 10 25 bulk wrapped per bag
₹44,000.00INR / Case of 500
COMPARE
Red 25 25 bulk wrapped per bag
₹24,700.00INR / Case of 200
COMPARE
Purple 50 25 bulk wrapped per bag
₹65,600.00INR / Case of 200
COMPARE
Blue 5 50 bulk wrapped per bag
₹14,600.00INR / Case of 200
COMPARE
Orange 10 50 bulk wrapped per bag
₹16,400.00INR / Case of 200
COMPARE
Orange 10 50 bulk wrapped per bag
₹23,300.00INR / Case of 200
COMPARE
Red 25 25 bulk wrapped per bag
₹15,700.00INR / Case of 100
Showing 1- 15 of 15