This product is now discontinued, View Related Products
Cole-Parmer StableTemp Laboratory Refrigerators
Built to meet the requirements of daily lab applications
  • Fully insulated chamber ensures minimal temperature fluctuation
  • Sliding wire shelves pull out fully for easy access to all samples
  • Thermal line recorder for easy tracking of temperature
Our complete line of refrigerators provides the reliability that is essential for your valuable samples in the 2 to 10ºC (35 to 50ºF) range. Performance is driven on all models by a high-efficiency evaporation system made with low corrosive materials and a fully insulated stainless steel chamber. An intuitive and easy-to-use control panel has VFD and touch-sensitive buttons for controlling all the essential functions.
Adjustable full-extension sliding shelves provide efficient and customizable use of interior space and easy access to stored samples. Built-in 50-mm diameter cable port allows use of external temperature probes. Bright fluorescent lighting (except 52412-00 to -02) for convenient viewing of samples.
Safety features include high/low temperature and overcurrent protection, open door alarm, and automatic fan stop for minimization of heat loss. Keypad lock function and built-in door lock with key permit access control.
Call For Price
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
COMPARE
60 120 5.5
Call For Price
COMPARE
60 230 5.5
Call For Price
COMPARE
50 230 5.5
Call For Price
COMPARE
60 120 10.7
Call For Price
COMPARE
60 230 10.7
Call For Price
COMPARE
50 230 10.7
Call For Price
COMPARE
60 120 21.3
Call For Price
COMPARE
60 230 21.3
Call For Price
COMPARE
50 230 21.3
Call For Price
COMPARE
60 120 28.4
Call For Price
COMPARE
60 230 28.4
Call For Price
COMPARE
50 230 28.4
Call For Price
COMPARE
60 120 46.9
Call For Price
COMPARE
60 230 46.9
Call For Price
COMPARE
50 230 46.9
Call For Price
Showing 1- 15 of 15