This product is now discontinued, View Related Products
Cole-Parmer StableTemp Economical Digital Utility Water Baths
Offering digital technology at analog prices
  • Easy-to-read LCD display shows your choice of °C or °F
  • Temperature can be recalled at the touch of a button — program up to 3 commonly used setpoints
  • Built-in timer notifies you when your process is complete
  • High-temperature alarm and safety shutdown features keep you and your bath safe and working
StableTemp utility baths combine precise temperature control and excellent temperature stability to provide you with reliable heating. These baths feature dual thermostats — a primary microprocessor-based temperature controller that provides ±0.1°C precision, and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer.
The stainless steel tank is seamless — no welds to leak or rust — and the rounded corners provide an easy-to-clean interior. Hinged polycarbonate lid allows condensation to flow back.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
115 2
Call For Price
COMPARE
230 2
Call For Price
COMPARE
115 5
Call For Price
COMPARE
230 5
Call For Price
COMPARE
115 10
Call For Price
COMPARE
230 10
Call For Price
COMPARE
115 20
Call For Price
COMPARE
230 20
Call For Price
COMPARE
115 28
Call For Price
COMPARE
230 28
Call For Price
Showing 1- 10 of 10