This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Heating Blocks for Vials
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Vial Blocks are single blocks and are designed for sample/serum and scintillation vials. Blocks are available for vial sizes from 12mm to 28mm in diameter.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mm)
Availability Pricing
COMPARE
20 12
Call For Price
COMPARE
20 15
Call For Price
COMPARE
15 16
Call For Price
COMPARE
12 17
Call For Price
COMPARE
12 19
Call For Price
COMPARE
9 21
Call For Price
COMPARE
8 23
Call For Price
COMPARE
8 25
Call For Price
COMPARE
6 28
Call For Price
Showing 1- 9 of 9