This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Heating Blocks for Test Tubes
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Test Tube Blocks are single blocks. Blocks are available for test tube sizes from 6mm to 35mm in diameter.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block - Tube / Vial Size Accepted (mm)
Availability Pricing
COMPARE
30 6
Call For Price
COMPARE
24 10
Call For Price
COMPARE
20 12
Call For Price
COMPARE
12 15
Call For Price
COMPARE
12 17
Call For Price
COMPARE
8 20
Call For Price
COMPARE
6 25
Call For Price
COMPARE
4 35
Call For Price
Showing 1- 8 of 8