This product is now discontinued, View Related Products
Cole-Parmer StableTemp Dry Block Heater, Heating Blocks for PCR Plates, Tubes, and Strips
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. The PCR Plate block has a thermometer well for measuring block temperature and is a double size block. Please note that the PCR Tube and PCR Strip blocks do not have a thermometer well and are single blocks. Tapered tube wells for 0.2mL tubes are spaced for easy access and removal.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Block Number Of Openings Block Type
Availability Pricing
COMPARE
1
Call For Price
COMPARE
10
Call For Price
COMPARE
64