Foxx Life Sciences Single-Use Sterile Vented Media Bottles, PC
Keep your system free of machining residue or bonding agents
  • Assemblies are gamma irradiated and made entirely with USP Class VI components
  • Manufactured in a certified ISO Class 7 cleanroom
These single-use media bottles with tubing and vent filters are perfect for fluid transfer and storage. Each cap features a platinum-cured silicone gasket for a leakproof system without any machining residue or bonding agents. Hose barb plug on tubing keeps line closed until ready for use. Use the pinch clamp to help control flow into the container or to close off the line. A 0.2 µm hydrophobic PTFE filter allows for ventilation while preventing media from escaping in the event of a spill. Rigid dip tube with notch provides easy filling and extraction of media.
192,000.00INR / Case of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size
Availability Pricing
Foxx Life Sciences Media Bottle, PC, 1000 mL; 10/Cs WW-06061-48 Mfr # 438-0006-FLS
COMPARE
1000 38-430
₹192,000.00INR / Case of 10