This product is now discontinued, View Related Products
Cole-Parmer Single-Channel Fixed-Volume Pipettors
Excellent accuracy and performance at an economical price
  • Individually calibrated in accordance with DIN 12650
Ergonomic, lightweight design with soft plunger movement minimizes hand fatigue. Pipettors feature streamlined, low-pressure tip ejector for easy access to narrow containers. They facilitate last drop dispensing and reverse pipetting technique with two-stage plunger operation for blow-out. Easy to calibrate and disassemble for routine servicing or autoclaving. Adjustable-volume pipets feature quick, easy click-stop volume settings.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy (%) Max Volume (mL) Max Volume (µL)
Availability Pricing
COMPARE
±0.30% 0.1 100
Call For Price
COMPARE
±0.30% 0.2 200
Call For Price
COMPARE
±0.30% 0.5 500
Call For Price
COMPARE
±0.30% 1 1000
Call For Price
COMPARE
±0.30% 5 5000