Cole-Parmer® Safety Goggles
For protection against chemical splash and other contaminants
  • Economical and comfortable goggle
  • 99.99% UV protection (UV-A, UV-B, UV-C)
  • Fits over most prescription glasses
  • Polycarbonate antiscratch lens with antifog coating
  • Ultrasonic-sealed indirect HDPE vent caps
  • 13mm Headband for user comfort
  • Class 1 high optical quality lenses
  • Dust proof, wind proof and splash protection
Certifications:
• CE certified
• EN166:2001 compliant
• ANSI Z87.1-2015 compliant
2,100.00 - ₹25,100.00INR / Box of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Color Description
Availability Pricing
COMPARE
Clear Clear Safety Goggles; 10 pcs/box
₹2,100.00INR / Box of 10
COMPARE
Clear Clear Safety Goggles, 10 pcs/box; 12 box/case
₹25,100.00INR / Case of 120