This product is now discontinued, View Related Products
Cole-Parmer Rotary Evaporator Systems with Motorized Lift
Convenient, quality evaporator systems at affordable prices
  • Small foorprint takes up minimal lab space
  • Motorized lift to easily raise and lower system
Rotary evaporators provide constant rotational speeds up to 280 rpm. Vertical glassware saves bench space and diagonal glassware keeps the profile low. Motorized lift features up and down buttons, and has an automatic raising function in the event of a power failure. Overall height is adjusted manually by a separate slide mechanism.
Glassware set includes 1-L pear-shaped evaporating flask, 1-L round-bottom receiving flask, and condenser. Select a model without a bath if you already have one, or choose between a model with a water bath for low boiling point applications and a water/oil bath for high boiling points.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Bath Type Condenser Type Power (VAC)
Availability Pricing
COMPARE
None Diagonal 115
Call For Price
COMPARE
Water Diagonal 115
Call For Price
COMPARE
Water & oil Diagonal 115
Call For Price
COMPARE
None Vertical 115
Call For Price
COMPARE
Water Vertical 115
Call For Price
COMPARE
Water & oil Vertical 115