Cole-Parmer® CH-800 Series Recirculating Chillers, –10 to 70°C
Maximize airflow and reduce maintenance with self-cleaning air filter
  • Continuous fluid level monitoring protects pump and prevents freezing
  • Heat up to 70°C, without additional options
  • Easy access to front fill reservoir port saves time and avoids spills
  • Noise reduction system creates quieter work environment
  • Fluid path stays clean without chemical additives
  • USB port allows for data logging
Available with either a turbine or positive displacement pump, these Cole-Parmer Polystat recirculating chillers feature a modulated refrigeration system designed to use less refrigerant, enhance temperature stability, extend compressor life, and allow you to migrate to natural refrigerants with little or no global warming effects. Heat up to 70°C, without additional options. Ideal for applications, such as lasers, that must be brought to a temperature above ambient before operation can begin.
Self-changing air filter system automatically changes the filter media at user-definable intervals or once a month for a two-year period helping to reduce downtime and maintenance. An easy-to-read large full-color touchscreen display offers a multilanguage interface and shows set temperature, actual fluid temperature, fluid pressure, fluid level, air filter status, and timer.
State-of-the-art capacitance liquid level sensor provides continuous monitoring to protect the pump from premature failure. A special noise reduction system makes the chillers extremely quiet while still maintaining optimal temperature control. A UV light biological growth inhibitor eliminates the need for chemical growth inhibitors and prevents algicides from being released into the waterways. Self-diagnostic system allows you to troubleshoot performance issues against data stored on the chiller the day it was manufactured.
Call For Price
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 20° C (watts) Power (VAC) Pump Type
Availability Pricing
COMPARE
700 240 Positive displacement
Call For Price
COMPARE
850 120 Positive displacement
Call For Price
COMPARE
700 240 Turbine
Call For Price
COMPARE
850 120 Turbine
Call For Price
COMPARE
1280 240 Turbine
Call For Price
COMPARE
600 120 Turbine
Call For Price
COMPARE
1280 240 Positive displacement
Call For Price
COMPARE
600 120 Positive displacement
Call For Price
COMPARE
1740 120 Turbine
Call For Price
COMPARE
1840 240 Turbine
Call For Price
COMPARE
1740 120 Positive displacement
Call For Price
COMPARE
1840 240 Positive displacement
Call For Price
COMPARE
290 208 / 240 Positive displacement
Call For Price
COMPARE
2650 240 Positive displacement
Call For Price
COMPARE
290 208 / 240 Turbine
Call For Price
COMPARE
2650 240 Turbine
Call For Price
Showing 1- 16 of 16