Cole-Parmer® Essentials PTFE Tissue Grinders (Plain tip plungers)
Plain tip plungers effectively homogenize a variety of tissue samples
  • Capable of reducing cellualar tissue to primary cells
  • Plunger head is pure PTFE with 6.5 mm stainless steel shaft
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass tubes are made of precision borosilicate glass. The pure PTFE plunger head is threaded onto a 6.5 mm stainless steel shaft. Grinding chamber clearance is 0.15 to 0.25 mm.
5,100.00 - ₹10,700.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Dispersion Type Overall Length (mm)
Availability Pricing
COMPARE
2 Fine 230
₹6,600.00INR / Each
COMPARE
5 Fine 235
₹5,100.00INR / Each
COMPARE
10 Fine 270
₹5,600.00INR / Each
COMPARE
15 Fine 270
₹6,900.00INR / Each
COMPARE
30 Fine 270
₹8,400.00INR / Each
COMPARE
50 Fine 270
₹10,700.00INR / Each