This product is now discontinued, View Related Products

Environmental Express® Preserved and Certified VOA Vials

Minimize hazardous chemical exposure with pre-filled vials
  • Level 1 cleaned, 40-mL vials include a Certificate of Analysis
  • Open-top, 0.125" septa caps are standard—choose from clear or amber glass
  • Meet EPA "Specifications and Guidance for Contaminant-Free Sample Containers"
After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.
11,200.00 - ₹28,600.00INR / Case of 72

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description Fill Tube Color
Availability Pricing
COMPARE
Clear VOA Vials, 40 mL, Open-Top Cap, 0.2 mL Hydrochloric Acid; 72/Cs 0.2 mL 1:1 hydrochloric acid (HCI) Clear
Call For Price
COMPARE
Amber VOA Vials, 40 mL, 0.2mL 1:1 HCL; 72/Cs 0.2mL 1:1 Hydrochloric acid Amber
Call For Price
COMPARE
Amber VOA Vials, 40 mL, Open-Top Cap, 0.3 mL 2% Sodium thiosulfate; 72/Cs 0.3 mL 2% Sodium thiosulfate Amber
Call For Price
COMPARE
Clear VOA Vials, 40 mL, Closed-Top, 25 mL Methanol; 20/Cs 25 mL Methanol Clear
Call For Price
COMPARE
Clear VOA Vials, 40 mL, Open-Top Cap, Stir Bar, 5 mL Sodium Bisulfate; 72/Cs 5 mL sodium bisulfate (NaHSO4) Clear
₹28,600.00INR / Case of 72
COMPARE
Clear VOA Vials, 40 mL, Open-Top Cap, 0.1 mL 25% Ascorbic Acid; 72/Cs 0.1 mL, 25% ascorbic acid (C6H806) Clear
₹11,200.00INR / Case of 72
COMPARE
Clear VOA Vials, 40 mL, Open-Top Cap, 1 mL Ascorbic Acid; 72/Cs 1.0 mL ascorbic acid (C6H806) Clear
₹16,700.00INR / Case of 72
COMPARE
Amber VOA Vials, 40 mL, Open-Top Cap, 0.5 mL 1:1 Sulfuric Acid; 72/Cs 0.5 mL, 1:1 sulfuric acid (H2SO4) Amber
Call For Price
COMPARE
Amber VOA Vials, 2% Sodium Thiosulfate; 72/Cs 2% sodium thiosulfate (Na2S2O3) Amber
₹14,100.00INR / Case of 72
COMPARE
Amber VOA Vials, 20 mL, Open-Top Cap, 1 mL MCAA buffer with Sodium thiosulfate; 144/Cs 1 mL MCAA buffer with Sodium thiosulfate Amber
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat