Cole-Parmer® Essentials Pre-Cleaned EPA Wide-Mouth Jars, Clear Glass
Certified clean for worry-free sampling
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined caps protect contents
  • Identify samples easier with included analysis labels (unattached)
These pre-cleaned, narrow-mouth jars are processed and certified to meet the requirements of the current US EPA “Specifications and Guidance for Contaminant-Free Sample Containers” document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyses.
6,500.00 - ₹13,700.00INR / Case of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
COMPARE
45 Glass 4 1/2
₹13,700.00INR / Case of 48
COMPARE
60 Glass 1 27/32
₹8,800.00INR / Case of 24
COMPARE
125 Glass 2 3/4
₹11,800.00INR / Case of 24
COMPARE
250 Glass 3 45/64
₹6,500.00INR / Case of 12
COMPARE
500 Glass 3 51/64
₹8,100.00INR / Case of 12
COMPARE
1000 Glass 6 51/64
₹10,400.00INR / Case of 12