Cole-Parmer® Essentials Pre-Cleaned EPA Narrow-Mouth Bottles, Amber Glass
Certified clean for worry-free sampling
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined caps protect contents
  • Identify samples easier with included analysis labels (unattached)
These pre-cleaned, narrow-mouth bottles are processed and certified to meet the requirements of the current US EPA “Specifications and Guidance for Contaminant-Free Sample Containers” document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyses.
5,600.00 - ₹12,400.00INR / Case of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 22-400 Glass
₹8,100.00INR / Case of 24
COMPARE
250 24-400 Glass
₹8,000.00INR / Case of 12
COMPARE
500 28-400 Glass
₹5,600.00INR / Case of 12
COMPARE
30 20-400 Glass
₹12,400.00INR / Case of 48
COMPARE
60 20-400 Glass
₹10,000.00INR / Case of 24