Cole-Parmer® Essentials Porcelain Gooch Filter Crucibles with Cover
Accepts filter paper and membranes
  • Autoclavable
Gooch filter crucibles are glazed inside and out, except for the outside bottom and rim. Bottom surface is perforated to accept filter papers and membranes. Use for determination of total suspended solids.
2,400.00 - ₹32,600.00INR / Pkg of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
30 36
₹32,600.00INR / Pkg of 24
COMPARE
140 55
₹28,100.00INR / Pkg of 6
COMPARE
15 29
₹6,300.00INR / Pkg of 6
COMPARE
30 36
₹8,300.00INR / Pkg of 6
COMPARE
50 40
₹16,000.00INR / Pkg of 6
COMPARE
140 55
₹2,400.00INR / Each