This product is now discontinued, View Related Products
Cole-Parmer PolyStat Refrigerated Tank
Reach below-ambient temperatures in an open tank!
This refrigerated deep drawn, seamless SS, tank is best used with an immersion circulator — select the circulator that best fits your temperature control needs and create your own constant temperature system. The bath alone does not have a temperature control device built in, and simply runs at maximum cooling capacity. Maximum cooling capacity is based largely on the ambient temperature of the bath and overall environment. An outlet can be found on the back of the bath for plugging in a heating device directly, reducing the need for multiple outlets to run the system.
Tank has a large 13-liter capacity and a working depth of 7-3/4". Order hollow balls to create a floating cover.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
150 0 , Refrigerated Tank, 13 L; 120VAC/60Hz
Call For Price
COMPARE
150 0 , Refrigerated Tank, 13 L; 240VAC/50Hz