This product is now discontinued, View Related Products

Cole-Parmer Polystat Heating Immersion Circulators

High-stability immersion circulators feature low-level sensor for temperature control up to 200°C
  • PID control with easy-to-use LCD display in three languages and up to five temperature setpoints
  • Pump connections for circulation to external equipment in open- or closed-loop applications
  • Resolution of 0.01° reads out in °C, °F, or °K
The immersion circulator is designed with an easy-to-read LCD in your choice of temperature units from Celsius/Farrenheit/or Kelvin. Programmable high/low/limit set points on easy-to-use PID temperature controller. Adjustable pump nozzle.
263,000.00 - ₹274,000.00INR / Each

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
100 115 Standard Immersion Circulator w/Clamp; 115V/60Hz 12120-01
Call For Price
COMPARE
100 230 Standard Immersion Circulator with Clamp; 230V/50Hz 12120-02
₹263,000.00INR / Each
COMPARE
150 115 Standard+ Immersion Circulator w/Clamp; 115V/60Hz 12120-03
Call For Price
COMPARE
150 230 Standard Plus Immersion Circulator with Clamp; 230V/50Hz 12120-04
₹265,000.00INR / Each
COMPARE
100 115 Standard Immersion Circulator w/Bridge; 115V/60Hz
Call For Price
COMPARE
100 230 Standard Immersion Circulator w/Bridge; 230V/50Hz
₹272,000.00INR / Each
COMPARE
150 115 Standard+ Immersion Circulator w/Bridge; 115V/60Hz
Call For Price
COMPARE
150 230 Standard+ Immersion Circulator w/Bridge; 230V/50Hz
₹274,000.00INR / Each
COMPARE
200 115 Advanced Immersion Circulator w/Bridge; 115V/60Hz
Call For Price
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat