Cole-Parmer® PolarSafe® Stainless Steel Liquid Nitrogen Refill Tanks
Self-pressurized LN2 tank makes LN2 discharge safe and simple
  • Durable welded stainless steel construction ensures long-lasting performance and reduced evaporation loss
  • Unique design utilizes pressure from LN2 to discharge LN2 to other containers
  • Use as a self-pressurized LN2 backup system for cryogenic freezers
  • 5 year vacuum warranty
The Cole-Parmer® PolarSafe® stainless steel liquid nitrogen (LN2) refill tanks feature a unique design ideal for cryopreservation applications. The tanks are self-pressurized utilizing pressure generated from the vaporization of small amounts of LN2 within the tank. This pressure allows for the discharge of LN2 to other containers. This unique design is more stable compared to traditional backup systems due to the reduction of intermediate pipe connections. These tanks also feature a lower working pressure, making them safer to use.
337,000.00 - ₹423,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Evaporation (Liters/day) OutsideDiameter (OD) (in) Volume (Liters)
Availability Pricing
COMPARE
0.75 17 29/32 30
₹337,000.00INR / Each
COMPARE
1 19 29/32 50
₹423,000.00INR / Each