This product is now discontinued, View Related Products
Cole-Parmer Pipette Tip Storage Boxes
Ideal for pipette tip protection and storage
  • Hinged lid provides fast, safe pipette tip access
  • Tip rack layout meets ANSI SLAS 1-2004
Each pipette tip storage box contains a hinged lid for easy one-hand opening, and a grid insert for quick organization. Each tip box can safely house a variety of pipette tip brands.
The transparent polycarbonate boxes featuring a 128-place grid insert, are designed for autoclaving tips at 121°C. They are 100% form stable, even after numerous autoclave cycles. Two sizes are available; one to accommodate 200 μL pipette tips and the other for 1000 μL pipette tips.
The polypropylene boxes are available in four sizes to accommodate either 20-, 200-, 300-, or 1000-µl pipette tips. Each box comes with a 96-place color coded grid insert.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Body Material Max Volume (µL) Description
Availability Pricing
COMPARE
PP 20 Pipette Tip Box, PP, with gray rack for 20 µl tips
Call For Price
COMPARE
PP 300 Pipette Tip Box, PP, with green rack for 300 µl tips
Call For Price
COMPARE
PP 200 Pipette Tip Box, PP, with yellow rack for 200 µl tips
Call For Price
COMPARE
PP 1000 Pipette Tip Box, PP, with blue rack for 1000 µl tips
Call For Price
COMPARE
PC 200 Pipette Tip Box, PC, with rack for 200 µl tips
Call For Price
COMPARE
PC 1000 Pipette Tip Box, PC, with rack for 1000 µl tips