This product is now discontinued, View Related Products
Cole-Parmer Pipette Tip Replacement Racks
Cleanroom produced for ready to use applications
  • Constructed of pure, high-quality polypropylene without additives
  • Certified RNase, DNase, and endotoxin free
  • Autoclavable at 121°C for 20 minutes
The tips are available in non-sterile and sterile rack options, as well as non-sterile and sterile stacked rack options. These are designed to give high levels of accuracy, precision, and reproducibility for challenging applications. The smooth surface of the tips minimizes binding of enzymes and nucleic acids to the walls. All tips are suitable for use with common market pipettes.
Call For Price
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Max Volume (µL) Package Of Sterile
Availability Pricing
COMPARE
20 10 refill racks of 96 each No
Call For Price
COMPARE
20 10 refill racks of 96 each No
Call For Price
COMPARE
50 10 refill racks of 96 each No
Call For Price
COMPARE
200 10 refill racks of 96 each No
Call For Price
COMPARE
300 10 refill racks of 96 each No
Call For Price
COMPARE
1000 10 refill racks of 96 each No
Call For Price
COMPARE
20 10 refill racks of 96 each Yes
Call For Price
COMPARE
20 10 refill racks of 96 each Yes
Call For Price
COMPARE
50 10 refill racks of 96 each Yes
Call For Price
COMPARE
200 10 refill racks of 96 each Yes
Call For Price
COMPARE
300 10 refill racks of 96 each Yes
Call For Price
COMPARE
1000 10 refill racks of 96 each Yes
Call For Price
COMPARE
20 2 x 5 stacked refill racks of 96 each No
Call For Price
COMPARE
200 2 x 5 stacked refill racks of 96 each No
Call For Price
COMPARE
1000 2 x 5 stacked refill racks of 96 each No
Call For Price
COMPARE
20 2 x 5 stacked refill racks of 96 each Yes
Call For Price
COMPARE
200 2 x 5 stacked refill racks of 96 each Yes
Call For Price
COMPARE
1000 2 x 5 stacked refill racks of 96 each Yes
Call For Price
Showing 1- 18 of 18