Cole-Parmer pH Electrodes with Interchangeable Cables
Save space and money by using one cable with multiple electrodes or vice versa
  • Cable cap rotates independently and will not bunch up
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
10,900.00 - ₹39,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH electrode, combination, rugged, glass body, interchangeble connection
₹20,400.00INR / Each
COMPARE
Interchangeable Cable pH Probe, Refillable/SJ/Glass; 154x12mm
₹20,300.00INR / Each
COMPARE
Interchangeable Cable Refillable SJ Glass pH Probe; 210x6mm
₹22,000.00INR / Each
COMPARE
Interchangeable Cable Refillable SJ Glass pH Probe; 329x8mm
₹39,000.00INR / Each
COMPARE
pH electrode, combination, rugged, epoxy body, interchangeable connection
₹10,900.00INR / Each
COMPARE
pH electrode, combination, spear tip, glass body, interchangeable connection
₹25,300.00INR / Each
COMPARE
Interchangeable Cable ORP Probe, Sealed/SJ/Epoxy; 155x12mm
₹20,900.00INR / Each
Showing 1- 7 of 7