Cole-Parmer® Perforated Insert Trays for Ultrasonic Cleaners
Achieve optimal cleaning results
  • Supports items that require direct cleaning
  • Prevents damage to transducers
Made of 304 stainless steel, these perforated trays can be used to support items in the tank that require direct cleaning. Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
21,800.00 - ₹59,400.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
COMPARE
304 SS Perforated Tray for 0.5 gal and SS 0.75 gal Cleaners
₹21,800.00INR / Each
COMPARE
304 SS Perforated Tray for 0.75 gal and SS 1 gal Cleaners
₹25,500.00INR / Each
COMPARE
304 SS Perforated Tray for 1.5 gal and SS 1.5 gal Cleaners
₹29,500.00INR / Each
COMPARE
304 SS Perforated Tray for 2.5 gal and SS 2.5/3.5 gal Cleaners
₹33,400.00INR / Each
COMPARE
304 SS Perforated Tray for 5.5 gal and SS 5.5 gal Cleaners
₹59,400.00INR / Each