This product is now discontinued, View Related Products
Cole-Parmer Orbital Shakers
Versatile units to meet your gentle shaking needs
  • Smooth orbital shaking action with 16-mm orbit — ideal for larger samples and multi-well plates
  • Built-in digital timer and variable speed control to 300 rpm
These orbital shakers provide a smooth, uniform circular motion with 16-mm orbit. Speed and time may be varied from 30 to 300 rpm, and 1 to 999 minutes—easily set and read on the digital display. Use in incubators, and environmental or cold chambers up to 104°F (40°C) and 80% humidity.
144,000.00 - ₹201,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Load (lbs) Power (VAC)
Availability Pricing
COMPARE
6.6 120
₹168,000.00INR / Each
COMPARE
6.6 230
₹144,000.00INR / Each
COMPARE
22 120
Call For Price
COMPARE
22 230
₹201,000.00INR / Each