Cole-Parmer® RH-400 Series Open Heated Circulating Bath Systems
Safely heat liquids with an intelligent display and simple controls
  • Backlit display prominently shows process and set values
  • Heating and circulating icons enhance user monitoring
  • Closed loop functionality provides greater temperature stability
  • On-screen prompts simplify functionality
These economical circulating baths are ideal for academic institutions, startups, and basic small-scale applications. External circulation through a closed loop provides greater stability at the point of use, while the tank material keeps the cost down. Economical temperature controller displays actual and set point temperatures simultaneously on backlit LCD. Safety features include overtemperature protection, high and low alarms, and low liquid level cutoff.
357,000.00 - ₹482,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
COMPARE
85 115 8
₹357,000.00INR / Each
COMPARE
85 230 8
₹357,000.00INR / Each
COMPARE
85 115 14
₹415,000.00INR / Each
COMPARE
85 230 14
₹415,000.00INR / Each
COMPARE
85 115 23
₹463,000.00INR / Each
COMPARE
135 120 6
₹382,000.00INR / Each
COMPARE
135 240 6
₹382,000.00INR / Each
COMPARE
135 120 10
₹401,000.00INR / Each
COMPARE
85 120 28
₹482,000.00INR / Each
COMPARE
85 240 28
₹482,000.00INR / Each
Showing 1- 10 of 10