Cole-Parmer Nylon Membrane Filters
Suitable for HPLC sample preparation
  • Inherently hydrophilc
  • Binds proteins, DNA, and RNA
Designed by impregnating a polyester web with nylon polymer, the membrane filters are very stong and heat resistant. Compatible with aqueous and alcoholic solutions and solvents.
38,100.00 - ₹106,000.00INR / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
25 Nylon 0.1
₹45,100.00INR / Pkg of 100
COMPARE
47 Nylon 0.1
₹58,300.00INR / Pkg of 100
Advantec Nylon Membrane Filters, 0.22 µm, 293 mm; 25/Pk WW-36230-52 Mfr # N022A293C
COMPARE
293 Nylon 0.22
₹106,000.00INR / Pkg of 25
Advantec Nylon Membrane Filters, 0.8 µm, 47 mm; 100/Pk WW-36230-54 Mfr # N080A047A
COMPARE
47 Nylon 0.8
₹59,800.00INR / Pkg of 100
Advantec Nylon Membrane Filters, 1.2 µm, 47 mm; 100/Pk WW-36230-58 Mfr # N120A047A
COMPARE
47 Nylon 1.2
₹52,500.00INR / Pkg of 100
Advantec Nylon Membrane Filters, 5.0 µm, 25 mm; 100/Pk WW-36230-60 Mfr # N500A025A
COMPARE
25 Nylon 5
₹38,100.00INR / Pkg of 100
COMPARE
47 Nylon 5.0
₹52,500.00INR / Pkg of 100
Advantec Nylon Membrane Filters, 5.0 µm, 90 mm; 25/Pk WW-36230-64 Mfr # N500A090C
COMPARE
90 Nylon 5
₹47,900.00INR / Pkg of 25
Showing 1- 8 of 8