Environmental Express® Non-Certified, Level 3, VOA Vials
Perfect for general use
  • Level 3 vials meet EPA standards but do not contain a certificate of analysis—ideal for every-day applications
  • Choose from open-top/septa caps or screw-top caps in 20-, 40-, or 60-mL capacities
  • Select amber glass for light-sensitive samples
Level 3 vials still conform to EPA "Specifications and Guidance for Contaminant-Free Sample Containers."
8,700.00 - ₹18,600.00INR / Case of 72
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Cap Type Volume (mL)
Availability Pricing
COMPARE
Clear VOA Vials, 20 mL, PTFE-Lined Cap; 72/Cs PTFE-lined screw-top 20
₹10,900.00INR / Case of 72
COMPARE
Amber VOA Vials, 40 mL, Open-Top Septa; 72/Cs Open-top/septa 40
₹8,700.00INR / Case of 72
COMPARE
Amber VOA Vials, 40 mL, Open-Top Septa, Stir Bar; 72/Cs Open-top/septa 40
₹18,600.00INR / Case of 72
COMPARE
Amber VOA Vials, 40 mL, PTFE-Lined Cap; 72/Cs PTFE-lined screw-top 40
₹13,000.00INR / Case of 72
COMPARE
Clear VOA Vials, 40 mL, Open-Top Septa; 72/Cs Open-top/septa 40
₹10,400.00INR / Case of 72
COMPARE
Clear VOA Vials, 40 mL, PTFE-Lined Cap; 72/Cs PTFE-lined screw-top 40
₹11,900.00INR / Case of 72