Cole-Parmer® ST-200 Series Multiposition Stirrers
Versatile—perform different mixing applications at the same time
  • Each stirring position is individually controlled
  • Capable of stirring solutions from 100 to 1500 rpm
  • Designed to keep spills away from the electronics
  • Solid ceramic top offers excellent corrosion and chemical resistance
These analog multiposition stirrers offer more versatility than single-position stirrers. Stirring positions are individually controlled so you can perform different mixing applications simultaneously. Each stirrer position can stir solutions from 100 to 1500 rpm. The solid ceramic top surface is corrosion and chemical resistant, is easy to clean, and measures 30.5 x 30.5 cm (12” x 12”).
323,000.00 - ₹452,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Stirring Volume per position (Liters) No. Of Stirring Positions Power (VAC)
Availability Pricing
COMPARE
0.8 5 120
₹323,000.00INR / Each
COMPARE
0.5 9 120
₹452,000.00INR / Each
COMPARE
0.8 5 230
₹323,000.00INR / Each
COMPARE
0.5 9 230
₹452,000.00INR / Each